Learn More
Embi Tec GE-3600: 1% Agarose TAE w/EtBr Landscape Gel,12+1 Well
SDP
Catalog No. NC1013872
Encompass
1% Agarose TAE w/EtBr Landscape Gel,12+1 WellEasy to UseJust open gel package and place gel with tray directly onto the RunOne Unit platform or similar size platform.ConvenientKeep stock on hand with a 7 Month Shelf Life. Store at 4-24ºC.
Catalog No. NC1013872 Supplier Embi Tec Supplier No. GG3600
1% Agarose TAE w/EtBr Landscape Gel,12+1 WellEasy to UseJust open gel package and place gel with tray directly onto the RunOne Unit platform or similar size platform.ConvenientKeep stock on hand with a 7 Month Shelf Life. Store at 4-24ºC.