Learn More
Embi Tec 1% AGAROSE TAE W/ETBR GEL 12+1
SDP
Catalog No. NC9139814
Encompass
1% Agarose TAE w/ EtBr, Landscape Gel 12 + 1 well, Load Volume: 30microliter, Optimal Separation: 600 to > 10000 bp, Dimensions: 11.5cm x 5.7cm, 10/BX, Gel Shipped in UVT Tray
Catalog No. NC9139814 Supplier Embi Tec Supplier No. GE3600
1% Agarose TAE w/ EtBr, Landscape Gel 12 + 1 well, Load Volume: 30microliter, Optimal Separation: 600 to > 10000 bp, Dimensions: 11.5cm x 5.7cm, 10/BX, Gel Shipped in UVT Tray