Learn More
Embi Tec GE-3730: 1% Agarose TAE w/EtBr Long Gel 2(12+1) Well
SDP
Catalog No. NC1128997
Encompass
1% Agarose TAE w/EtBr Long Gel 2(12+1) WellEasy to UseJust open gel package and place gel with tray directly onto the RunOne Unit platform or similar size platform.ConvenientKeep stock on hand with a 7 Month Shelf Life. Store at 4-24° C.
Catalog No. NC1128997 Supplier Embi Tec Supplier No. GE3730
1% Agarose TAE w/EtBr Long Gel 2(12+1) WellEasy to UseJust open gel package and place gel with tray directly onto the RunOne Unit platform or similar size platform.ConvenientKeep stock on hand with a 7 Month Shelf Life. Store at 4-24° C.