Learn More
Embi Tec GE-3640: 1% Agarose TAE w/EtBr Long Gel 4(24+1) Well
SDP
Catalog No. NC1070870
Encompass
1% Agarose TAE w/EtBr Long Gel 4(24+1) WellEasy to UseJust open gel package and place gel with tray directly onto the RunOne Unit platform or similar size platform.ConvenientKeep stock on hand with a 7 Month Shelf Life. Store at 4-24° C.
Catalog No. NC1070870 Supplier Embi Tec Supplier No. GG3640
1% Agarose TAE w/EtBr Long Gel 4(24+1) WellEasy to UseJust open gel package and place gel with tray directly onto the RunOne Unit platform or similar size platform.ConvenientKeep stock on hand with a 7 Month Shelf Life. Store at 4-24° C.