Learn More
Embi Tec 1% Agarose TAE with GelGreen Portrait Prep Gel
SDP
Catalog No. NC1037091
Encompass
1% Agarose TAE w/GelGreen Portrait Prep Gel, 5+1 Well
Catalog No. NC1037091 Supplier Embi Tec Supplier No. GG5100
1% Agarose TAE w/GelGreen Portrait Prep Gel, 5+1 Well