Learn More
Embi Tec 1% AGOROSE GEL (12X) 10/BOX
SDP
Catalog No. NC9564874
Encompass
1% Agarose TAE w/EtBr Portrait Gel, 12 well 10/box
Catalog No. NC9564874 Supplier Embi Tec Supplier No. GE3700
1% Agarose TAE w/EtBr Portrait Gel, 12 well 10/box