Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Allyl-3,5-dimethylpyrazole 97.0+%, TCI America™
SDP

Catalog No. A16795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16795G Supplier TCI America Supplier No. A16795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13369-74-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00034407
InChI Key NXAGTQRRYZTDKV-UHFFFAOYSA-N
Synonym 1-allyl-3,5-dimethylpyrazole, 1-allyl-3,5-dimethyl-1h-pyrazole, pyrazole, 1-allyl-3,5-dimethyl, 3,5-dimethyl-1-prop-2-en-1-yl pyrazole, acmc-209bta, 5-23-05-00127 beilstein handbook reference, pyrazole, 3,5-dimethyl-1-allyl, 1-allyl-3,5-dimethyl-1h-pyrazole #, 3,5-dimethyl-1-prop-2-en-1-yl-1h-pyrazole, 1h-pyrazole,3,5-dimethyl-1-2-propen-1-yl
PubChem CID 25930
IUPAC Name 3,5-dimethyl-1-prop-2-enylpyrazole
SMILES CC1=CC(=NN1CC=C)C

Specifications

Color Yellow
Boiling Point 192°C
Quantity 5 g
Formula Weight 136.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Allyl-3,5-dimethylpyrazole
RTECSNumber UQ4975000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.