Alfa Aesar™ 1-Allyl-3-methylimidazolium chloride, 98%

Manufacturer: Alfa Aesar™ H2695218

Quantity 50g

 View more versions of this product

Catalog No. AAH2695218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 65039-10-3
    Molecular Formula C7H11ClN2
    Formula Weight 158.63
    MDL Number MFCD07784450
    Synonym AMIMCl
    Chemical Name or Material 1-Allyl-3-methylimidazolium chloride

    Specifications

    Formula Weight 158.63
    Molecular Formula C7H11ClN2
    MDL Number MFCD07784450
    CAS 65039-10-3
    Melting Point 49° to 51°C
    Synonym AMIMCl
    Quantity 50g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1-Allyl-3-methylimidazolium chloride
    Assay 98%

    Description

    Applications
    1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.

    Notes
    Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit