Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Allylhydantoin 98.0+%, TCI America™
SDP

Catalog No. A114425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A114425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A114425G Supplier TCI America Supplier No. A114425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3366-93-6
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00142580
InChI Key KPDTTZWHFZUVCL-UHFFFAOYSA-N
PubChem CID 22020924
IUPAC Name 1-prop-2-enylimidazolidine-2,4-dione
SMILES C=CCN1CC(=O)NC1=O

Specifications

Melting Point 97°C
Color White
Quantity 25 g
Formula Weight 140.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Allylhydantoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.