Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Allylisatin 98.0+%, TCI America™
SDP

Catalog No. A29431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29431G Supplier TCI America Supplier No. A29431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 830-74-0
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD00224218
InChI Key ZWNYDPBLEDGGQD-UHFFFAOYSA-N
Synonym 1-Allylindole-2,3-dione, 1-Allyl-2,3-indolinedione
PubChem CID 1713563
IUPAC Name 1-(prop-2-en-1-yl)-2,3-dihydro-1H-indole-2,3-dione
SMILES C=CCN1C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 86°C
Color Red-Yellow
Boiling Point 145°C
Quantity 1 g
Formula Weight 187.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Allylisatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.