Learn More
Cayman Chemical 1-AlnInchlamydocIn 2.5mg
SDP
Supplier:  Cayman Chemical 281022.5 MG
Encompass_Preferred
An HDAC inhibitor; reduces total HDAC activity in HeLa cell lysates; reduces proliferation of MIA PaCa-2, PANC-1, and hTERT-HPNE cells (GI50s = 5.3, 14, and 2.0 nM, respectively)
Catalog No. 50-293-8135