Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-1-cyclopentanecarboxylic acid, 97%

Catalog No. AC103470100
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103470100 10 g Glass bottle
AC103470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103470100 Supplier Thermo Scientific Chemicals Supplier No. 103470100
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 52-52-8 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-52-8
MDL Number MFCD00001381
InChI Key NILQLFBWTXNUOE-UHFFFAOYSA-N
Synonym cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
PubChem CID 2901
ChEBI CHEBI:40547
IUPAC Name 1-aminocyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H11NO2
Linear Formula H2NC5H8CO2H
Quantity 10 g
Beilstein 14, I, 526
Merck Index 15, 2727
Molecular Weight (g/mol) 129.159
Formula Weight 129.16
Percent Purity ≥96.0%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 1-Amino-1-cyclopentanecarboxylic acid, 97%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-144-6
RTECSNumber GY2625000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.