Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-2-butanol 98.0+%, TCI America™
SDP

Catalog No. A08235ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08235ML 5 mL
A082325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. A08235ML Supplier TCI America Supplier No. A08235ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13552-21-1
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00059196
InChI Key KODLUXHSIZOKTG-UHFFFAOYSA-N
PubChem CID 26107
IUPAC Name 1-aminobutan-2-ol
SMILES CCC(CN)O

Specifications

Color Colorless
Boiling Point 169°C
Quantity 5 mL
UN Number 2735
Formula Weight 89.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Amino-2-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.