Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-2-methylnaphthalene Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A133325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A133325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A133325G Supplier TCI America Supplier No. A133325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111180-78-0
Molecular Formula C11H12ClN
Molecular Weight (g/mol) 193.674
MDL Number MFCD00136475
InChI Key PEBKGVSIRFSIGW-UHFFFAOYSA-N
Synonym 2-Methyl-1-naphthylamine Hydrochloride
PubChem CID 24213295
IUPAC Name 2-methylnaphthalen-1-amine;hydrochloride
SMILES CC1=C(C2=CC=CC=C2C=C1)N.Cl

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 25 g
Formula Weight 193.67
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-2-methylnaphthalene Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.