Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-4-(2-hydroxyethyl)piperazine, 96%, Thermo Scientific™

Catalog No. p-7052643
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55134MD 250 mg
AAH5513406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH55134MD Supplier Thermo Scientific Chemicals Supplier No. H55134MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3973-70-4
Molecular Formula C6H15N3O
Molecular Weight (g/mol) 145.21
MDL Number MFCD00006155
InChI Key XAOHWSFGKCJOGT-UHFFFAOYSA-N
PubChem CID 77597
IUPAC Name 2-(4-aminopiperazin-1-yl)ethan-1-ol
SMILES NN1CCN(CCO)CC1

Specifications

Melting Point 105°C to 107°C
Color Brown to White
Quantity 250 mg
Synonym 1-amino-4-2-hydroxyethyl piperazine, 2-4-aminopiperazin-1-yl ethanol, 1-piperazineethanol, 4-amino, 4-amino-1-piperazineethanol, 1-piperazineethanol,4-amino, 2-4-aminopiperazinyl ethan-1-ol, 2-4-amino-piperazin-1-yl-ethanol, 1-amino-4-2-hydroxyethyl-piperazine
Formula Weight 145.2
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1-Amino-4-(2-hydroxyethyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.