Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Amino-4-chloronaphthalene

Catalog No. A464045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A464045G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A464045G Supplier Sigma Aldrich Supplier No. A464045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5-methoxy-1,2,3-triazine is a 1,2,3-triazine derivative bearing electron-donating substituent
  • It participates in the inverse electron demand diels-alder reactions
  • 5-methoxy-1,2,3-triazine readily reacts with amidines
  • It can be prepared from 4-methoxypyrazole.5-methoxy-1,2,3-triazine may be used in the synthesis of nitrogen-containing heterocyclic compounds.cycloadditions of 1,2,3-triazines bearing c5-electron donating substituents: robust pyrimidine synthesis

Specifications

Melting Point 98°C to 100°C (lit.)
Molecular Formula C10H8ClN
Linear Formula H2NC10H6Cl
Quantity 100 mg
MDL Number MFCD00004024
Synonym 4-Chloro-1-naphthylamine
Molecular Weight (g/mol) 177.63
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.