Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-4-hydroxyanthraquinone 98.0+%, TCI America™
SDP

Catalog No. A03145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A03145G Supplier TCI America Supplier No. A03145G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 116-85-8
Molecular Formula C14H9NO3
Molecular Weight (g/mol) 239.23
MDL Number MFCD00001223
InChI Key AQXYVFBSOOBBQV-UHFFFAOYSA-N
Synonym 1-amino-4-hydroxyanthraquinone, disperse red 15, calcosyn pink b, celutate pink b, nacelan pink b, amacel pink b, celutate pink bn, celutate pink by, cibacete red 3b, disperse red 2s
PubChem CID 8323
IUPAC Name 1-amino-4-hydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N

Specifications

Melting Point 215°C
Quantity 5 g
Formula Weight 239.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-4-hydroxyanthraquinone
EINECSNumber (4)-0716
RTECSNumber CB5600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.