Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-4-methylpiperazine, 99%

Catalog No. AC251391000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC251391000 100 g Glass bottle
AC251390250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC251391000 Supplier Thermo Scientific Chemicals Supplier No. 251391000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 6928-85-4 | C5H13N3 | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6928-85-4
Molecular Formula C5H13N3
Molecular Weight (g/mol) 115.18
MDL Number MFCD00006154
InChI Key RJWLLQWLBMJCFD-UHFFFAOYSA-N
Synonym 1-amino-4-methylpiperazine, 1-piperazinamine, 4-methyl, 4-methyl-1-piperazinamine, 4-methyl-piperazin-1-ylamine, 1-methyl-4-aminopiperazine, 4-amino-1-methylpiperazine, 4-methylpiperazinylamine, 1-methyl-4-aminopierazine, pubchem8577, n-amino-4-methylpiperazine
PubChem CID 81349
IUPAC Name 4-methylpiperazin-1-amine
SMILES CN1CCN(CC1)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.9500g/mL
Boiling Point 172.0°C to 175.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4835 to 1.4855
Quantity 100 g
Beilstein 23, 376
Specific Gravity 0.95
Solubility Information Solubility in water: miscible
Formula Weight 115.18
Percent Purity 99%
Physical Form Viscous Solution
Chemical Name or Material 1-Amino-4-methylpiperazine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
EINECSNumber 230-053-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.