Learn More
Description
- Methyl thiosalicylate may be benefecial in preventing mercury-induced toxicity.methyl thiosalicylate may be used:
- In the synthesis of thioxanthone, an efficient enantioselective organocatalyst for the intramolecular [2+2] photocycloaddition reaction
- As anionic bidentate ligand, in the preparation of α2+1α type complexes of [(99m)tc]-tricarbonyltechnetium(i) and [(188)re]-tricarbonylrhenium(i)
- In the synthesis of a series of benzisothiazolone derivatives
- In the synthesis of a new fused benzoheterocyclic compound, [1]benzothieno[3,2-b]furan
Specifications
Specifications
| Molecular Formula | C10H13N |
| Linear Formula | C10H13N |
| MDL Number | MFCD07373907 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 147.22 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.