Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-8-naphthol-4-sulfonic Acid 90.0+%, TCI America™
SDP

Catalog No. A036925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A036925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A036925G Supplier TCI America Supplier No. A036925G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83-64-7
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.25
MDL Number MFCD00003992
InChI Key LRDIEHDJWYRVPT-UHFFFAOYSA-N
Synonym s acid, 1-amino-8-naphthol-4-sulfonic acid, chicago acid s, 4-amino-5-hydroxy-1-naphthalenesulfonic acid, kyselina s, 1-naphthalenesulfonic acid, 4-amino-5-hydroxy, aminonaphtholsulfonic acid s, kyselina s czech, aminonaphthol sulfonic acid s, kyselina 1-amino-8-naftol-4-sulfonova
PubChem CID 6752
IUPAC Name 4-amino-5-hydroxynaphthalene-1-sulfonic acid
SMILES NC1=C2C(O)=CC=CC2=C(C=C1)S(O)(=O)=O

Specifications

Color Black-Gray
Quantity 25 g
Formula Weight 239.25
Percent Purity ≥90.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-8-naphthol-4-sulfonic Acid
EINECSNumber (4)-0513
RTECSNumber QK1295000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.