Learn More
Medchemexpress LLC 1-Amino-N-(4-(6-methyl-1,2,4,5-tetrazin-3-yl)benzyl)-3,6,9,12-tetraoxapentadecan-15-amide HCl | 2656478-51-0 | 95.0% | 484.98 g/mol | C21H33ClN6O5 | 5 MG
SDP
Catalog No. 5000262262
Encompass_Preferred
Me-Tet-PEG4-NH2 hydrochloride is a methyltetrazine-functionalized PEG4 amine supplied as the hydrochloride salt. It is used as a click-chemistry reagent and linker for bioconjugation, enabling rapid inverse electron demand Diels-Alder (IEDDA) reactions with trans-cyclooctenes and facilitating PROTAC and antibody-drug conjugate assembly.
  • Provides a methyltetrazine handle for fast IEDDA click reactions.
  • Contains a primary amine for standard coupling chemistries.
  • Incorporates a PEG4 spacer to improve aqueous solubility and reduce steric hindrance.
  • Supplied as hydrochloride salt for enhanced stability and handling.
  • Suitable for use in PROTAC and antibody-drug conjugate linker assembly.
  • Typical purity ~95.0% by LCMS as reported on the certificate of analysis.
Catalog No. 50-002-62262 Supplier Medchemexpress LLC Supplier No. HY1335055MG
Me-Tet-PEG4-NH2 hydrochloride is a methyltetrazine-functionalized PEG4 amine supplied as the hydrochloride salt. It is used as a click-chemistry reagent and linker for bioconjugation, enabling rapid inverse electron demand Diels-Alder (IEDDA) reactions with trans-cyclooctenes and facilitating PROTAC and antibody-drug conjugate assembly.
  • Provides a methyltetrazine handle for fast IEDDA click reactions.
  • Contains a primary amine for standard coupling chemistries.
  • Incorporates a PEG4 spacer to improve aqueous solubility and reduce steric hindrance.
  • Supplied as hydrochloride salt for enhanced stability and handling.
  • Suitable for use in PROTAC and antibody-drug conjugate linker assembly.
  • Typical purity ~95.0% by LCMS as reported on the certificate of analysis.