Learn More
Enzo Life Sciences 1-Aminobenzotriazole (100mg). CAS: 1614-12-6
SDP
Catalog No. 502010640
Encompass_Preferred
Suicide substrate for cytochrome P-450. Inhibits oxidative drug metabolism in vivo. Blocks ω-oxidation of arachidonic acid but not epoxidation. Purity: ≥99% (GC). Solubility: Soluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0640 Supplier Enzo Life Sciences Supplier No. BMLEI2590100
Add to Cart
Add to Cart
Suicide substrate for cytochrome P-450. Inhibits oxidative drug metabolism in vivo. Blocks ω-oxidation of arachidonic acid but not epoxidation. Purity: ≥99% (GC). Solubility: Soluble in water. Long Term Storage: Ambient.