Learn More
Medchemexpress LLC 1-aminobenzotriazole | 1614-12-6 | MFCD00132902 | 99.9% | 134.14 g/mol | C6H6N4 | 1 ML
SDP
Catalog No. 5000244564
Encompass_Preferred
1-Aminobenzotriazole (ABT) is a nonspecific, irreversible inhibitor of cytochrome P450 enzymes. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research use in biochemical and cell-based assays. The compound has molecular formula C6H6N4 and a molecular weight of 134.14 g/mol.
  • Nonspecific, irreversible inhibitor of cytochrome P450 enzymes
  • Supplied as a 10 mM solution in DMSO for convenient assay preparation
  • 1 mL volume suitable for small-scale or single-use experiments
  • Molecular formula C6H6N4 and molecular weight 134.14 g/mol for reference
  • High purity (~99.9%) to minimize experimental impurities
Catalog No. 50-002-44564 Supplier Medchemexpress LLC Supplier No. HY10338910MM 1ML
1-Aminobenzotriazole (ABT) is a nonspecific, irreversible inhibitor of cytochrome P450 enzymes. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research use in biochemical and cell-based assays. The compound has molecular formula C6H6N4 and a molecular weight of 134.14 g/mol.
  • Nonspecific, irreversible inhibitor of cytochrome P450 enzymes
  • Supplied as a 10 mM solution in DMSO for convenient assay preparation
  • 1 mL volume suitable for small-scale or single-use experiments
  • Molecular formula C6H6N4 and molecular weight 134.14 g/mol for reference
  • High purity (~99.9%) to minimize experimental impurities