Learn More
Medchemexpress LLC 1-aminobenzotriazole | 1614-12-6 | MFCD00132902 | 99.9% | 134.14 g/mol | C6H6N4 | 100 MG
SDP
Catalog No. 5000245176
Encompass_Preferred
1-Aminobenzotriazole is a nonspecific, irreversible inhibitor of cytochrome P450 enzymes used as a research reagent to block P450-mediated metabolism in biochemical and pharmacological studies. It is supplied as a white to off-white powder with high reported purity for laboratory use.
  • Irreversible cytochrome P450 inhibitor suitable for in vitro and in vivo research.
  • High purity: 99.88% (manufacturer reported).
  • Molecular formula C6H6N4 and molecular weight 134.14 g/mol.
  • Available in multiple laboratory pack sizes and DMSO solution formats.
  • Intended for research use only; not for human therapeutic or diagnostic use.
Catalog No. 50-002-45176 Supplier Medchemexpress LLC Supplier No. HY103389100MG
1-Aminobenzotriazole is a nonspecific, irreversible inhibitor of cytochrome P450 enzymes used as a research reagent to block P450-mediated metabolism in biochemical and pharmacological studies. It is supplied as a white to off-white powder with high reported purity for laboratory use.
  • Irreversible cytochrome P450 inhibitor suitable for in vitro and in vivo research.
  • High purity: 99.88% (manufacturer reported).
  • Molecular formula C6H6N4 and molecular weight 134.14 g/mol.
  • Available in multiple laboratory pack sizes and DMSO solution formats.
  • Intended for research use only; not for human therapeutic or diagnostic use.