Learn More
Medchemexpress LLC 1-aminobenzotriazole | 1614-12-6 | MFCD00132902 | 99.9% | 134.14 g/mol | C6H6N4 | 200 MG
SDP
Catalog No. 5000244974
Encompass_Preferred
1-aminobenzotriazole is a nonspecific, irreversible inhibitor of cytochrome P450 (CYP) enzymes used in laboratory research to block metabolic activity and study drug metabolism.
  • Nonspecific, irreversible inhibitor of cytochrome P450 enzymes.
  • Used to inhibit in vitro and in vivo CYP-mediated metabolism studies.
  • Available in small research pack sizes, including a 200 mg solid.
  • High chemical purity (approximately 99.9%).
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-44974 Supplier Medchemexpress LLC Supplier No. HY103389200MG
1-aminobenzotriazole is a nonspecific, irreversible inhibitor of cytochrome P450 (CYP) enzymes used in laboratory research to block metabolic activity and study drug metabolism.
  • Nonspecific, irreversible inhibitor of cytochrome P450 enzymes.
  • Used to inhibit in vitro and in vivo CYP-mediated metabolism studies.
  • Available in small research pack sizes, including a 200 mg solid.
  • High chemical purity (approximately 99.9%).
  • For research use only; not for human or veterinary use.