Learn More
Medchemexpress LLC HY-103389 500mg Medchemexpress, 1-Aminobenzotriazole ABT;3-Aminobenzotriazole CAS:1614-12-6 Purity:98%
SDP
Catalog No. 501637151
Encompass_Preferred
Medchemexpress,HY-103389 500mg 1-Aminobenzotriazole ABT;3-Aminobenzotriazole CAS:1614-12-6 Formula:C6H6N4 1-Aminobenzotriazole is a nonspecific and irreversible inhibitor of cytochrome P450 (P450). Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7151 Supplier Medchemexpress LLC Supplier No. HY103389500MG
Medchemexpress,HY-103389 500mg 1-Aminobenzotriazole ABT;3-Aminobenzotriazole CAS:1614-12-6 Formula:C6H6N4 1-Aminobenzotriazole is a nonspecific and irreversible inhibitor of cytochrome P450 (P450). Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.