Learn More
Selleck Chemical LLC 1-Aminobenzotriazole
SDP
Catalog No. 5000002715
Encompass_Preferred
1-Aminobenzotriazole (ABT 3-Aminobenzotriazole 1-Benzotriazolylamine NSC 114498 NSC 656987) is a nonselective and irreversible inhibitor of cytochrome P450 (CYP) enzymes 1-Aminobenzotriazole is also a substrate and inhibitor of N-acetyltransferase (NAT)
Catalog No. 50-000-02715 Supplier Selleck Chemical LLC Supplier No. S688625MG
1-Aminobenzotriazole (ABT 3-Aminobenzotriazole 1-Benzotriazolylamine NSC 114498 NSC 656987) is a nonselective and irreversible inhibitor of cytochrome P450 (CYP) enzymes 1-Aminobenzotriazole is also a substrate and inhibitor of N-acetyltransferase (NAT)