Learn More
Cayman Chemical 1-AmInobenzotrIazole 250mg
SDP
Catalog No. 502973318
Encompass_Preferred
A non-specific CYP inhibitor; acts as a suicide substrate, producing maximal destruction of hepatic and renal microsome CYP protein in vitro at 10 mM; effective in vivo, significantly reducing CYP content in both liver and kidney in rats within two hours
Catalog No. 50-297-3318 Supplier Cayman Chemical Supplier No. 15252250 MG
A non-specific CYP inhibitor; acts as a suicide substrate, producing maximal destruction of hepatic and renal microsome CYP protein in vitro at 10 mM; effective in vivo, significantly reducing CYP content in both liver and kidney in rats within two hours