Learn More
TARGETMOL CHEMICALS INC 1-Aminobenzotriazole 500MG
SDP
Catalog No. 502879926
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 1-Aminobenzotriazole (ABT) (ABT) is a non-specific cytochrome P450 (CYP) inhibitor. Purity 98.77%
Catalog No. 50-287-9926 Supplier TARGETMOL CHEMICALS INC Supplier No. T5953500MG
Add to Cart
Add to Cart
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 1-Aminobenzotriazole (ABT) (ABT) is a non-specific cytochrome P450 (CYP) inhibitor. Purity 98.77%