Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminocyclobutanecarboxylic Acid Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A20325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20325G Supplier TCI America Supplier No. A20325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98071-16-0
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD08752285
InChI Key HBTVGNDTGRUBQO-UHFFFAOYSA-N
Synonym 1-aminocyclobutanecarboxylic acid hydrochloride, 1-amino-1-cyclobutanecarboxylic acid hydrochloride, 1-aminocyclobutane-1-carboxylic acid hydrochloride, cyclobutanecarboxylic acid, 1-amino-, hydrochloride, 1-amino-cyclobutanecarboxylic acid hydrochloride, 1-amino-cyclobutanecarboxylic acid hcl, 1-amino cyclobutane carboxylic acid hcl, 1-aminocyclobutanecarboxylic acid hcl, h-ac4c-oh*hcl, 1-amino
PubChem CID 11321048
IUPAC Name 1-aminocyclobutane-1-carboxylic acid hydrochloride
SMILES Cl.NC1(CCC1)C(O)=O

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 5 g
Formula Weight 151.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminocyclobutanecarboxylic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.