Learn More
Specifications
Specifications
| Color | White |
| Quantity | 250 mg |
| Synonym | 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid |
| PubChem CID | 535 |
| Formula Weight | 101.1 |
| Physical Form | Solid |
| Chemical Name or Material | 1-Aminocyclopropane-1-Carboxylic Acid |
Safety and Handling
| Recommended Storage | Store at 0°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.