Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminocyclopropane-1-Carboxylic Acid, MP Biomedicals™

Catalog No. ICN15034891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15034891 1 g
ICN15034880 100 mg
ICN15034883 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15034891 Supplier MP Biomedicals, Inc Supplier No. 0215034891
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22059-21-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00009944
InChI Key PAJPWUMXBYXFCZ-UHFFFAOYSA-N
ChEBI CHEBI:18053
IUPAC Name 1-aminocyclopropane-1-carboxylic acid
SMILES NC1(CC1)C(O)=O

Specifications

Color White
Quantity 1 g
Synonym 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
PubChem CID 535
Formula Weight 101.1
Physical Form Solid
Chemical Name or Material 1-Aminocyclopropane-1-Carboxylic Acid
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.