Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminocyclopropanecarboxylic acid hydrochloride, 97%

Catalog No. AC429731000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429731000 100 mg Glass bottle
AC429735000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429731000 Supplier Thermo Scientific Chemicals Supplier No. 429731000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 68781-13-5 | C4H7NO2·ClH | 137.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68781-13-5
Molecular Formula C4H7NO2·ClH
Molecular Weight (g/mol) 137.57
MDL Number MFCD00012545
InChI Key WTZRHZRMLYNBRM-UHFFFAOYSA-N
Synonym 1-aminocyclopropanecarboxylic acid hydrochloride, 1-aminocyclopropane-1-carboxylic acid hydrochloride, 1-amino-cyclopropane-1-carboxylic acid hydrochloride, 1-aminocyclopropanecarboxylic acid hcl, cyclopropanecarboxylic acid, 1-amino-, hydrochloride, 1-aminocyclopropanecarboxylic acid, chloride, acpc-hcl, pubchem13872, ksc352q0j, 1-amino-cyclopropane-1-carboxylic acid, hcl
PubChem CID 11957445
IUPAC Name 1-aminocyclopropane-1-carboxylic acid;hydrochloride
SMILES C1CC1(C(=O)O)N.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 221.6°C to 225.0°C
Color White
Elemental Analysis 33.52 to 36.31% C, 5.63 to 6.09% H, 9.77 to 10.59% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 137.57
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminocyclopropanecarboxylic acid hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.