Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Aminohydantoin | 6301-02-6 | 115.09 | 100 MG
SDP

Catalog No. 5000402141
Encompass_Preferred

1-Aminohydantoin is a Furagin derivative with a molecular weight of 115.09 and a chemical formula of C3H5N3O2. Its CAS number is 6301-02-6. This product is intended for research use only and is not for human use.

  • For research use only, not intended for human use.
  • Molecular weight of 115.09.
  • Chemical formula of C3H5N3O2.
  • CAS number 6301-02-6.
  • Ships at room temperature in continental US.
  • Store under recommended conditions in the Certificate of Analysis.

Catalog No. 50-004-02141 Supplier Medchemexpress LLC Supplier No. HY23506100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Aminohydantoin is a Furagin derivative with a molecular weight of 115.09 and a chemical formula of C3H5N3O2. Its CAS number is 6301-02-6. This product is intended for research use only and is not for human use.

  • For research use only, not intended for human use.
  • Molecular weight of 115.09.
  • Chemical formula of C3H5N3O2.
  • CAS number 6301-02-6.
  • Ships at room temperature in continental US.
  • Store under recommended conditions in the Certificate of Analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.