Learn More
Medchemexpress LLC 1-aminohydantoin hydrochloride | 2827-56-7 | MFCD02181047 | ≥98.0% | 151.55 | C3H6ClN3O2 | 500mg
SDP
Catalog No. 5000268235
Encompass_Preferred
1-Aminohydantoin hydrochloride (Standard) is the analytical standard of 1-Aminohydantoin hydrochloride (HY-Y0469) This product is intended for research and analytical applications 1-Aminohydantoin hydrochloride is a major metabolite of Nitrofurantoin (HY-A0090) in animal tissues and can be used as a standard for the determination of residues of veterinary agents in meat milk et al 1-Aminohydantoin hydrochloride covalently binds to tissue proteins and is released from the tissues under slightly acidic conditions and derivatized with 2-nitrobenzaldehyde to form nitrophenyl derivatives of 1-Aminohydantoin hydrochloride before detection[1][2]
Catalog No. 50-002-68235 Supplier Medchemexpress LLC Supplier No. HYY0469R500MG
1-Aminohydantoin hydrochloride (Standard) is the analytical standard of 1-Aminohydantoin hydrochloride (HY-Y0469) This product is intended for research and analytical applications 1-Aminohydantoin hydrochloride is a major metabolite of Nitrofurantoin (HY-A0090) in animal tissues and can be used as a standard for the determination of residues of veterinary agents in meat milk et al 1-Aminohydantoin hydrochloride covalently binds to tissue proteins and is released from the tissues under slightly acidic conditions and derivatized with 2-nitrobenzaldehyde to form nitrophenyl derivatives of 1-Aminohydantoin hydrochloride before detection[1][2]