Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminoisoquinoline 98.0+%, TCI America™
SDP

Catalog No. A28175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28175G 5 g
A28171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28175G Supplier TCI America Supplier No. A28175G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1532-84-9
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00024137
InChI Key OSILBMSORKFRTB-UHFFFAOYSA-N
PubChem CID 73742
IUPAC Name isoquinolin-1-amine
SMILES NC1=C2C=CC=CC2=CC=N1

Specifications

Melting Point 119°C
Boiling Point 166°C
Quantity 5 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminoisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.