Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(aminomethyl)cyclopropanecarboxylic acid

Catalog No. CDS015451 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS015451 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS015451 Supplier Sigma Aldrich Supplier No. CDS015451100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-allylcyclohexanone is a α,α-unsaturated ketone. selective oxidation of 2-allylcyclohexanone by benzonitrile-hydrogen peroxide has been reported. asymmetric synthesis of 2-alkylcyclohexanones by alkylation of cyclohexanone enamines (iii) of L-proline ester derivatives has been described. oxidation of 2-allylcyclohexanone by 90% hydrogen peroxide catalyzed by arsonated polystyrene has been studied.2-allylcyclohexanone may be used in the synthesis of the following:
  • Bicyclo[3.3.1]non-2-en-9-one
  • R-(-)-epilachnene, an antipode of the defensive droplets from the mexican bean beetle, epilachna varivestis

Specifications

Molecular Formula C5H9NO2
Linear Formula C5H9NO2
Quantity 5 g
MDL Number MFCD06212872
Molecular Weight (g/mol) 115.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.