Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminopentadecane 95.0+%, TCI America™
SDP

Catalog No. A07635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07635G 5 g
A076310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07635G Supplier TCI America Supplier No. A07635G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2570-26-5
Molecular Formula C15H33N
Molecular Weight (g/mol) 227.436
MDL Number MFCD00038195
InChI Key JPZYXGPCHFZBHO-UHFFFAOYSA-N
Synonym pentadecylamine, 1-aminopentadecane, n-pentadecylamine, 1-pentadecanamine, 1-pentadecylamine, pentadecane, 1-amino, unii-2f7n79m9vh, acmc-209gly, 4-04-00-00817 beilstein handbook reference
PubChem CID 17386
ChEBI CHEBI:87551
IUPAC Name pentadecan-1-amine
SMILES CCCCCCCCCCCCCCCN

Specifications

Melting Point 37°C
Color White
Quantity 5 g
UN Number 2923
Formula Weight 227.44
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminopentadecane
EINECSNumber (2)-0133
RTECSNumber RZ2120000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.