Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-aminopropan-2-ol | 78-96-6 | MFCD00008139 | 100.0% | 75.11 g/mol | C3H9NO | 25 G
SDP

Catalog No. 5000304197
Encompass_Preferred

1-Aminopropan-2-ol (monoisopropanolamine) is a small amino alcohol used as an endogenous metabolite and biochemical research reagent. It is involved in microbial amino alcohol metabolism and is provided as a high-purity material for laboratory and analytical workflows that require well-characterized standards.

  • High purity for analytical and research use.
  • Molecular weight 75.11 g/mol.
  • Chemical formula C3H9NO.
  • Useful as an endogenous metabolite standard in metabolic studies.
  • Supplied with datasheet and safety documentation.

Catalog No. 50-003-04197 Supplier Medchemexpress LLC Supplier No. 25GHYW015969
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Aminopropan-2-ol (monoisopropanolamine) is a small amino alcohol used as an endogenous metabolite and biochemical research reagent. It is involved in microbial amino alcohol metabolism and is provided as a high-purity material for laboratory and analytical workflows that require well-characterized standards.

  • High purity for analytical and research use.
  • Molecular weight 75.11 g/mol.
  • Chemical formula C3H9NO.
  • Useful as an endogenous metabolite standard in metabolic studies.
  • Supplied with datasheet and safety documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.