Learn More
Medchemexpress LLC 1-aminopropan-2-ol | 78-96-6 | MFCD00008139 | 75.11 g/mol | C3H9NO | 10 G
SDP
Catalog No. 5000301002
Encompass_Preferred
1-Aminopropan-2-ol (monoisopropanolamine) is an amino alcohol used as a research reagent and endogenous metabolite reference. It is involved in microbial amino alcohol metabolism and is supplied for analytical and laboratory applications. Handle and store according to solvent and temperature recommendations; intended for research use only.
  • Endogenous metabolite reference for metabolic and biochemical studies.
  • Provides solubility guidance for DMSO (200 mg/mL) and ready stock preparation volumes.
  • Stable when stored sealed and protected from moisture; in solvent, store -80°C for up to 6 months or -20°C for 1 month.
  • Available in multiple packaging sizes, including 10 g and larger quantities.
  • Suitable for analytical, biochemical, and microbial metabolism research applications.
Catalog No. 50-003-01002 Supplier Medchemexpress LLC Supplier No. HYW01596910G
1-Aminopropan-2-ol (monoisopropanolamine) is an amino alcohol used as a research reagent and endogenous metabolite reference. It is involved in microbial amino alcohol metabolism and is supplied for analytical and laboratory applications. Handle and store according to solvent and temperature recommendations; intended for research use only.
  • Endogenous metabolite reference for metabolic and biochemical studies.
  • Provides solubility guidance for DMSO (200 mg/mL) and ready stock preparation volumes.
  • Stable when stored sealed and protected from moisture; in solvent, store -80°C for up to 6 months or -20°C for 1 month.
  • Available in multiple packaging sizes, including 10 g and larger quantities.
  • Suitable for analytical, biochemical, and microbial metabolism research applications.