Learn More
Medchemexpress LLC 1-aminopropan-2-ol (monoisopropanolamine) | 78-96-6 | MFCD00008139 | 100.0% | 75.11 | C3H9NO | 50 G
SDP
Catalog No. 5000299671
Encompass_Preferred
1-Aminopropan-2-ol (monoisopropanolamine) is a low-molecular-weight amino alcohol used as a biochemical reagent and reference standard for metabolic studies, reagent formulation, and analytical applications. Supplied as a high-purity material, it is suitable for laboratory research and method development.
  • High purity for analytical and research use.
  • Small, polar amino alcohol suitable for metabolic studies and reagent synthesis.
  • Soluble in DMSO at high concentration; may require sonication.
  • Stable when stored sealed at 4°C; stock solutions stable at -80°C (6 months) or -20°C (1 month).
  • Offered in small laboratory pack sizes for research workflows.
Catalog No. 50-002-99671 Supplier Medchemexpress LLC Supplier No. HYW01596950G
1-Aminopropan-2-ol (monoisopropanolamine) is a low-molecular-weight amino alcohol used as a biochemical reagent and reference standard for metabolic studies, reagent formulation, and analytical applications. Supplied as a high-purity material, it is suitable for laboratory research and method development.
  • High purity for analytical and research use.
  • Small, polar amino alcohol suitable for metabolic studies and reagent synthesis.
  • Soluble in DMSO at high concentration; may require sonication.
  • Stable when stored sealed at 4°C; stock solutions stable at -80°C (6 months) or -20°C (1 month).
  • Offered in small laboratory pack sizes for research workflows.