Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminotridecane 98.0+%, TCI America™
SDP

Catalog No. A076225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A076225G 25 g
A076210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A076225G Supplier TCI America Supplier No. A076225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2869-34-3
Molecular Formula C13H29N
Molecular Weight (g/mol) 199.382
MDL Number MFCD00008156
InChI Key ABVVEAHYODGCLZ-UHFFFAOYSA-N
Synonym 1-aminotridecane, 1-tridecanamine, tridecylamine, n-tridecylamine, monotridecylamine, tridecanamine, 1-tridecylamine, unii-5s0kq9wz2d, 5s0kq9wz2d, tridecyl-amine
PubChem CID 40761
IUPAC Name tridecan-1-amine
SMILES CCCCCCCCCCCCCN

Specifications

Color White-Yellow
Boiling Point 275°C
Quantity 25 g
UN Number 3259
Formula Weight 199.38
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 1-Aminotridecane
EINECSNumber (2)-0133
RTECSNumber YD3011650
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.