Learn More
Medchemexpress LLC HY-134145 5mg Medchemexpress, 1-(Anilinocarbonyl)proline CAS:73096-22-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1341455MG
Encompass_Preferred
Medchemexpress, HY-134145 5mg 1-(Anilinocarbonyl)proline CAS:73096-22-7 1-(Anilinocarbonyl)proline can be used to identify dual action probes in a cell model of Huntington. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1357