Learn More
Cayman Chemical 1-ArachIdonyl Glycerol 1mg
SDP
Catalog No. 502893357
Encompass_Preferred
An endogenous CB receptor ligand that is 10-100 times less potent than 2-AG in ligand binding affinity and agonist activity at the CB1 receptor; 2-AG undergoes rapid isomerization to 1-AG, which makes it a frequent contaminant in synthetic 2-AG preparations, markedly reducing their cannabinergic potency
Catalog No. 50-289-3357 Supplier Cayman Chemical Supplier No. 621501 MG
An endogenous CB receptor ligand that is 10-100 times less potent than 2-AG in ligand binding affinity and agonist activity at the CB1 receptor; 2-AG undergoes rapid isomerization to 1-AG, which makes it a frequent contaminant in synthetic 2-AG preparations, markedly reducing their cannabinergic potency