Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aza-12-crown 4-Ether 98.0+%, TCI America™
SDP

Catalog No. A1552250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1552250MG 250 mg
A15521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1552250MG Supplier TCI America Supplier No. A1552250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41775-76-2
Molecular Formula C8H18NO3
Molecular Weight (g/mol) 176.24
MDL Number MFCD00134269
InChI Key QNSRHBOZQLXYNV-UHFFFAOYSA-O
PubChem CID 545817
IUPAC Name 1,4,7-trioxa-10-azacyclododecan-10-ium
SMILES C1COCCOCCOCC[NH2+]1

Specifications

Melting Point 58°C
Quantity 250 mg
Synonym 1-aza-12-crown 4-ether, aza-12-crown-4, acmc-209spc, 1-aza-12-crown4-ether, 1-aza-4,7,10-trioxacyclododecane, 1,4,7-trioxa-10-aza cyclododecane, 1-aza-12-crown
Formula Weight 175.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aza-12-crown 4-Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.