Learn More
Selleck Chemical LLC 1-Azakenpaullone
SDP
Catalog No. 5000002876
Encompass_Preferred
1-Azakenpaullone (1-Akp) is a potent and selective GSK-3 inhibitor with IC50 of 18 nM 100-fold selectivity over CDK1/cyclin B and CDK5/p25
Catalog No. 50-000-02876 Supplier Selleck Chemical LLC Supplier No. S71932MG
1-Azakenpaullone (1-Akp) is a potent and selective GSK-3 inhibitor with IC50 of 18 nM 100-fold selectivity over CDK1/cyclin B and CDK5/p25