Learn More
Enzo Life Sciences 1-Azakenpaullone (1mg). CAS: 676596-65-9
SDP
Catalog No. 502008554
Encompass_Preferred
Analog of kenpaullone (BML-EI310) acting as a potent and ATP-competitive inhibitor of GSK-3β (IC50=18nM). Displays ~100-200 fold greater selectivity on GSK-3β than over CDK1/cyclin B and CDK5/p25 (IC50=2.0µM and 4.2µM, respectively). Alternative name: 9-Bromo-7,12-dihydropyrido[3',2':2,3]azepino[4,5-b]indol-6(5H)-one, 1-AKP. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in DMSO; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8554 Supplier Enzo Life Sciences Supplier No. ALX270430M001
Analog of kenpaullone (BML-EI310) acting as a potent and ATP-competitive inhibitor of GSK-3β (IC50=18nM). Displays ~100-200 fold greater selectivity on GSK-3β than over CDK1/cyclin B and CDK5/p25 (IC50=2.0µM and 4.2µM, respectively). Alternative name: 9-Bromo-7,12-dihydropyrido[3',2':2,3]azepino[4,5-b]indol-6(5H)-one, 1-AKP. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in DMSO; insoluble in water. Long Term Storage: -20°C.