Learn More
Selleck Chemical LLC 1-Azakenpaullone 5mg 676596-65-9
SDP
Supplier:  Selleck Chemical LLC S71935MG
Encompass_Preferred
1-Azakenpaullone is a potent and selective GSK-3? inhibitor with IC50 of 18 nM, >100-fold selectivity over CDK1/cyclin B and CDK5/p25. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4506