Learn More
AdipoGen 1-Azakenpaullone
SDP
Catalog No. 502562384
Encompass_Preferred
Chemical. CAS 676596-65-9. Formula C15H10BrN3O. MW 328.2. Potent and ATP-competitive GSK-3beta glycogen synthase kinase-3beta inhibitor IC50 = 18 nM.
Catalog No. 50-256-2384 Supplier AdipoGen Supplier No. AGCR10035M005
Chemical. CAS 676596-65-9. Formula C15H10BrN3O. MW 328.2. Potent and ATP-competitive GSK-3beta glycogen synthase kinase-3beta inhibitor IC50 = 18 nM.