Learn More
STEMCELL Technologies 1-Azakenpaullone, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 74252
Encompass
1-Azakenpaullone is a potent and highly selective inhibitor of glycogen synthase kinase 3β (IC₅₀ = 18 nM) and is highly preferential to GSK3β versus other cyclin-dependent kinases (CDKs) including CDK/cyclin B and CDK5/p25 (IC₅₀ = 2 µM and 4.2 µM, respectively; Kunick et al.).MAINTENANCE AND SELF-RENEWAL · In combination with ID-8 (Catalog #72502) and FK506 (Catalog #74152), supports human pluripotent stem cells in long-term propagation, maintaining pluripotency, and capacity to differentiate (Yasuda et al.).
Catalog No. 50-197-6205